Skip to Content
Merck
All Photos(1)

Key Documents

A8054

Sigma-Aldrich

D(−)-2-Amino-5-phosphonopentanoic acid

NMDA receptor antagonist

Synonym(s):

D(−)-AP-5, D(−)-APV, D-2-Amino-5-phosphonovaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12NO5P
CAS Number:
Molecular Weight:
197.13
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

optical purity

optical purity: ≥90% (HPLC, Marfey′s reagent)

technique(s)

ligand binding assay: suitable

color

white

mp

245-246 °C

SMILES string

N[C@H](CCCP(O)(O)=O)C(O)=O

InChI

1S/C5H12NO5P/c6-4(5(7)8)2-1-3-12(9,10)11/h4H,1-3,6H2,(H,7,8)(H2,9,10,11)/t4-/m1/s1

InChI key

VOROEQBFPPIACJ-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D(-)-2-Amino-5-phosphonopentanoic acid has been used to pretreat BNST (bed nucleus of the stria terminalis) in order to block the induction of anxiety by ORX-A (orexin-A) into the BNST.

Biochem/physiol Actions

Anticonvulsant; potent and selective N-methyl-D-aspartate (NMDA) receptor antagonist; active enantiomer of 2-amino-5-phosphonopentanoic acid.
NMDA antagonists have muscle relaxation property and are known to offer neuron protection against brain damage induced by ischemia. 2-Amino-5-phosphonopentanoic acid has been proved to block the iontophoresis induced depolarization of NMDA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synaptic Plasticity and Learning: Selective Impairment of Learning in
Rats and Blockade of Long-Term Potentiation in viva by the
IV-Methyl-D-Aspartate Receptor Antagonist AP5
R. G. M. Morris
The Journal of Neuroscience, 9(9), 3040-3057 (1989)
Paola Conti et al.
European journal of medicinal chemistry, 42(8), 1059-1068 (2007-03-10)
Twelve novel conformationally constrained homologues of glutamic acid have been synthesized and pharmacologically characterized at ionotropic glutamate receptors (iGluRs). Synthesis of the target compounds involved 1,3-dipolar cycloaddition of nitrile oxides to suitable dipolarophiles. The structure to the compounds has been
Orexin-A Induces Anxiety-like Behavior through Interactions with Glutamatergic Receptors in the Bed Nucleus of the Stria Terminalis of Rats
Elizabeth A Lungwitz
Physiology & Behavior, 107(5), 726-732 (2012)
Excitatory amino acid transmitters.
J C Watkins et al.
Annual review of pharmacology and toxicology, 21, 165-204 (1981-01-01)
Rachel E Field et al.
Neuron, 106(5), 842-854 (2020-03-28)
Excitation in neural circuits must be carefully controlled by inhibition to regulate information processing and network excitability. During development, cortical inhibitory and excitatory inputs are initially mismatched but become co-tuned or balanced with experience. However, little is known about how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service