Skip to Content
Merck

A7856

Acipimox

≥99% (TLC)

Synonym(s):

2-Carboxy-5-methylpyrazine 4-oxide, 5-Methylpyrazinecarboxylic acid 4-oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
MDL number:
UNSPSC Code:
12352205
EC Number:
256-928-3
NACRES:
NA.79
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (TLC)

form

powder

color

off-white to faint yellow

mp

177-180 °C

storage temp.

2-8°C

SMILES string

[n+]1(c(cncc1C(=O)O)C)[O-]

InChI

1S/C6H6N2O3/c1-4-2-7-3-5(6(9)10)8(4)11/h2-3H,1H3,(H,9,10)

InChI key

DNRXJHATQULEHC-UHFFFAOYSA-N

Biochem/physiol Actions

Niacin-derived, vasodilator studied for its lipid-lowering effect.
Acipimox, also known as olbemox, is a nicotinic acid analog. It functions as an anti-lipolytic drug and vasodilator. Acipimox may be used in various metabolic studies involving insulin and ghrelin. It lowers total cholesterol and total triglycerides, which helps in the treatment of hyperlipidemia.


Still not finding the right product?

Explore all of our products under Acipimox


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library