Skip to Content
Merck
All Photos(2)

Key Documents

A7502

Sigma-Aldrich

DL-Alanine

≥99% (HPLC)

Synonym(s):

(±)-2-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720250
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

mp

289 °C (dec.) (lit.)

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

CC(N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)

InChI key

QNAYBMKLOCPYGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Alanine, a racemic mixture of D- and L-alanine, is used as a model reagent for development of amino acid racemic resolution techniques. DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatjana Braun et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(30), 9340-9345 (2015-07-01)
The prokaryotic origins of the actin cytoskeleton have been firmly established, but it has become clear that the bacterial actins form a wide variety of different filaments, different both from each other and from eukaryotic F-actin. We have used electron
Randolph M Baral et al.
Journal of feline medicine and surgery, 17(8), 653-666 (2014-10-10)
In-house analysers are commonplace in small animal practices but cannot be calibrated by the operator; therefore, any bias in the generated plasma analyte values cannot be corrected. Guidelines such as grading of renal disease and published reference intervals (RIs) in
Janine Rohrbach et al.
Antiviral therapy, 19(2), 149-159 (2013-09-17)
We investigated changes in biomarkers of liver disease in HIV-HCV-coinfected individuals during successful combination antiretroviral therapy (cART) compared to changes in biomarker levels during untreated HIV infection and to HIV-monoinfected individuals. Non-invasive biomarkers of liver disease (hyaluronic acid [HYA], aspartate
Andrea Schlegel et al.
Annals of surgery, 260(5), 839-846 (2014-11-08)
To develop a reproducible animal model mimicking a novel 2-staged hepatectomy (ALPPS: Associating Liver Partition and Portal Vein Ligation for Staged Hepatectomy) and explore the underlying mechanisms. ALPPS combines portal vein ligation (PVL) with liver transection (step I), followed by
Gad Cotter et al.
European journal of heart failure, 16(12), 1362-1371 (2014-11-06)
Worsening heart failure (WHF) in the first 7 days after an admission for acute HF (AHF) has been proposed as a therapeutic target in several recent AHF studies and was a co-primary endpoint of the VERITAS studies. Patients were randomized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service