A7377
D-Alanine
≥98% (HPLC)
Synonym(s):
(R)-2-Aminopropionic acid
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
CH3CH(NH2)CO2H
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720249
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
D-Alanine, ≥98% (HPLC)
Assay
≥98% (HPLC)
form
powder
color
white
mp
291 °C (dec.) (lit.)
solubility
H2O: soluble
application(s)
detection
peptide synthesis
SMILES string
C[C@@H](N)C(O)=O
InChI
1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1
InChI key
QNAYBMKLOCPYGJ-UWTATZPHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Alanine is a non essential amino acid.
Application
D-Alanine has been used:
- as an amine donor for ω-transaminase mediated (R)-amination
- as a substrate for the treatment of D-amino acid oxidase (DAAO )-transfected cells
- to measure the heat capacity of L-alanine single crystal
Biochem/physiol Actions
D-Alanine is the non-proteinogenic form of alanine which is used in bacterial cell wall biosynthesis. D-Alanine is used to study the specificity and kinetics of alanine racemase(s) (alaR).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D-Amino Acids: Physiology, Metabolism, and Application (2016)
Amit Priyadarshi et al.
Biochimica et biophysica acta, 1794(7), 1030-1040 (2009-03-31)
Alanine racemase (AlaR) is a bacterial enzyme that belongs to the fold-type III group of pyridoxal 5'-phosphate (PLP)-dependent enzymes. AlaR catalyzes the interconversion between L- and D-alanine, which is important for peptidoglycan biosynthesis. This enzyme is common in prokaryotes, but
A system for omega-transaminase mediated (R)-amination using L-alanine as an amine donor
Richter N, et al.
Green Chemistry, 17(5), 2952-2958 (2015)
Srinivas Suda et al.
Journal of bacteriology, 194(3), 708-714 (2011-11-30)
Ltnα and Ltnβ are individual components of the two-peptide lantibiotic lacticin 3147 and are unusual in that, although ribosomally synthesized, they contain d-amino acids. These result from the dehydration of l-serine to dehydroalanine by LtnM and subsequent stereospecific hydrogenation to
Hookang Im et al.
BMC microbiology, 11, 116-116 (2011-05-27)
Streptococcus pneumoniae is a globally important pathogen. The Gram-positive diplococcus is a leading cause of pneumonia, otitis media, bacteremia, and meningitis, and antibiotic resistant strains have become increasingly common over recent years. Alanine racemase is a ubiquitous enzyme among bacteria
Chromatograms
application for HPLCapplication for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service