Skip to Content
Merck

A7094

L-Asparagine monohydrate

≥98.0%, suitable for cell culture, BioReagent, non-animal source

Synonym(s):

Asparagine monohydrate, (S)-(+)-2-Aminosuccinamic acid, (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2COCH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
150.13
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
200-735-9
MDL number:
Beilstein/REAXYS Number:
1723527
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Asparagine monohydrate, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%

biological source

non-animal source

Quality Segment

product line

BioReagent

assay

≥98.0%

form

powder

optical activity

[α]20/D 33.0 to 36.3 °, c = 10 in 6 M HCl

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

233-235 °C (lit.)

solubility

H2O: 20 mg/mL

cation traces

Fe: ≤10 ppm, heavy metals (as Pb): ≤10 ppm, <0.1% (Ammonia (NH3))

application(s)

advanced drug delivery

functional group

amide, amine, carboxylic acid

storage temp.

room temp

SMILES string

[H]O[H].N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1

InChI key

RBMGJIZCEWRQES-DKWTVANSSA-N

General description

L-Asparagine is a naturally occurring amino acid found in various proteins. It is used in laboratory research for various applications, including cell culture, protein biochemistry, and nitrogen metabolism studies. Its solubility in water and role as a source of asparagine make it a versatile tool for investigating biological processes and chemical synthesis. Further, L-Asparagine residues in proteins can be crucial for protein stability, folding, and interactions.

Application

L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine. L-Asparagine, a proteinogenic amino acid, from non-animal source is used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It has also been used as a component of LHC basal media for the culture of human airway epithelial cells.

Biochem/physiol Actions

The amino acid L-asparagine is a structural analog of L-aspartic acid, where the side chain of the carboxylic acid moiety is amidated to give a terminal amine group. This renders L-asparagine neutral at physiological pH. The amide group of asparagine is derived from glutamine, in the reaction of aspartate and glutamine in the presence of ATP to yield asparagine and glutamate. In vivo, asparagine is hydrolyzed to aspartate and NH4+ by asparaginase. Asparagine is also an important amino acid in glycopeptide bonds, via N-glycosyl linkages to the sugar rings.

Features and Benefits

  • Suitable for Cell Culture and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.


Still not finding the right product?

Explore all of our products under L-Asparagine monohydrate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library