Skip to Content
Merck

A7003

Alginic acid from brown algae

powder

Synonym(s):

Alginic acid from Macrocystis pyrifera (kelp), Mixed polymer of mannuronic and guluronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H8O6)n
CAS Number:
UNSPSC Code:
12171500
EC Number:
232-680-1
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

technique(s)

titration: suitable

color

white to light yellow

pH range

1.5-3.5

mp

300 °C

solubility

H2O: insoluble, but swells, aqueous base: soluble

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

General description

Alginic acid is a high molecular weight linear polymer isolated from seaweed. It is a linear β-1→4-linked polymer of n-mannuronic and L-guluronic acid. It is an acid polysaccharide present in the extracellular matrix of brown algae. It is reported to inhibit the reverse transcriptase activity of HIV. Structural investigation of alginic acid, a marine algal polysaccharide poly-α-L-guluronic acid, has been reported. Anti-inflammatory effect of alginic acid, isolated from the brown algae Sargassum wightii, in type II collagen induced arthritic rats has been reported. Pyrolysis of alginic acid using py-GC/MS over a wide temperature range (200-800°C) and thermogravimetric analysis (TGA) has been reported. It was reported to cause the biocorrosion of thin films of copper on germanium internal reflection elements (IREs).

Application

Alginic acid is suitable reagent used to study the in-vitro and in-vivo mitogenic activity of alginates. It is suitable for use in the spectrophotometric determination of transparent exopolymer particles by dye-binding assay.

Other Notes

A straight-chain, hydrophilic, colloidal, polyuronic acid composed primarily of anhydro-β-D-mannuronic acid residues with 1→4 linkage.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library