Skip to Content
Merck
All Photos(1)

Key Documents

A6986

Sigma-Aldrich

Acetyl-CoA Carboxylase 1 human

recombinant, expressed in Sf9 cells

Synonym(s):

ACAC, ACACA, ACC, ACC1, ACCA, acetyl-CoA carboxylase alpha

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.26

recombinant

expressed in Sf9 cells

Quality Level

form

liquid

specific activity

≥20 units/μg protein

NCBI accession no.

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ACACA(31)

General description

Acetyl-CoA Carboxylase 1 (ACACA) is mapped to human chromosome 17q12. It is majorly expressed in liver and adipose tissue. ACACA has biotin carboxylase (BC), biotin carboxyl carrier protein (BCCP) and carboxyl transferase (CT) domains and an additional interaction domain. (BT) A central domain region (CD) connects the BC and CT domains. ACACA is a key regulator of energy balance. The ACACA catalysis is the rate-limiting step in the synthesis of malonyl-CoA and regulation of free fatty acid oxidation. Elevated levels of ACACA contributes to obesity and tumor progression.
Formulation: Solution in Tris-HCl (pH 8), NaCl, Glycerol

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Acetyl-CoA Carboxylase (ACC) regulates the metabolism of fatty acids. This enzyme catalzes the formation of Malonyl CoA through the irreversible carboxylation of acetyl CoA. There are two main isoforms of Acetyl-CoA carboxylase expressed in mammals, Acetyl-CoA carboxylase 1 (ACACA) and Acetyl-CoA carboxylase 2 (ACACB). ACACA has broad tissue distribution but is enriched in tissues critical for fatty acid sythesis such as adipose tissue. ACACB is enriched in tissues such as skeletal muscle and heart that are critical for fatty acid oxidation.

The Acetyl-CoA Carboxylase enzymes are activated by citrate, glutamate, and dicarboxylic acids and negatively regulated by long and short chain fatty acyl CoAs. Acetyl-CoA Carboxylase 1 is essential for breast cancer and prostrate cancer cell survival. Because of thier roles in fatty acid metabolism and oxidation, ACACA and ACACB are therapeutic targets for treating obesity and metabolic syndrome disorders.

Physical properties

α transcript variant 1, C-terminal histidine-tagged 270 kDa protein containing amino acids 1-2383 (end)

Unit Definition

One unit will cause the conversion of 1 picomole of ATP to ADP per minute at pH 7.4 at 30 °C

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural basis for regulation of human acetyl-CoA carboxylase
Hunkeler M, et al.
Nature, 558, 470-474 (2018)
Regulation of acetyl CoA carboxylase and carnitine palmitoyl transferase-1 in rat adipocytes
Zang Y, et al.
Obesity Research, 13(9), 1530-1539 (2005)
Morgan D Fullerton et al.
Nature medicine, 19(12), 1649-1654 (2013-11-05)
The obesity epidemic has led to an increased incidence of nonalcoholic fatty liver disease (NAFLD) and type 2 diabetes. AMP-activated protein kinase (Ampk) regulates energy homeostasis and is activated by cellular stress, hormones and the widely prescribed type 2 diabetes
Graeme J Gowans et al.
Cell metabolism, 18(4), 556-566 (2013-10-08)
While allosteric activation of AMPK is triggered only by AMP, binding of both ADP and AMP has been reported to promote phosphorylation and inhibit dephosphorylation at Thr172. Because cellular concentrations of ADP and ATP are higher than AMP, it has
Karen Moreau et al.
The Journal of biological chemistry, 281(6), 3172-3181 (2005-12-06)
Germ line alterations in BRCA1 (breast cancer susceptibility gene 1) are associated with an increased susceptibility to breast and ovarian cancer. BRCA1 acts as a scaffold protein implicated in multiple cellular functions, such as transcription, DNA repair, and ubiquitination. However

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service