Skip to Content
Merck
All Photos(1)

Key Documents

A6751

Sigma-Aldrich

N-Acetyl-L-tyrosine ethyl ester monohydrate

≥98%, suitable for ligand binding assays

Synonym(s):

ATEE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO4 · H2O
CAS Number:
Molecular Weight:
269.29
Beilstein:
2217900
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-Acetyl-L-tyrosine ethyl ester monohydrate,

Assay

≥98%

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

CCOC(=O)[C@H](Cc1ccc(O)cc1)NC(C)=O

InChI

1S/C13H17NO4/c1-3-18-13(17)12(14-9(2)15)8-10-4-6-11(16)7-5-10/h4-7,12,16H,3,8H2,1-2H3,(H,14,15)/t12-/m0/s1

InChI key

SKAWDTAMLOJQNK-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-L-tyrosine ethyl ester monohydrate has been used:
  • as a substrate in transesterification reactions with 1 M propan-1-ol catalyzed by Carlsberg protease protein-coated microcrystals (PCMC)
  • to test the performance of subtilisin Carlsberg protein-coated microcrystals (PCMC)
  • as a substrate for cross-linked crystals (CLECs) of subtilisin activity screening

Biochem/physiol Actions

N-Acetyl-L-tyrosine ethyl ester is an N-terminal and C-terminal protected L-tyrosine that is used in crosslinking studies and as a substrate for the detection, differentiation and/or characterization various proteases and esterases.

Substrates

Substrate for chymotrypsin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sandra M Arriaga et al.
Clinical biochemistry, 42(9), 919-921 (2009-01-20)
To evaluate if unconjugated bilirubin (UB) inhibits C1 esterase activity. Esterase activity was evaluated by C1-mediated hydrolysis of N-acetyl-L-tyrosine ethyl ester, and binding of UB to C1r and C1s was assessed by dot-blot analysis. UB inhibited C1 enzymatic activity. C1r
Stability of protein-coated microcrystals in organic solvents
Kreiner M, et al.
Journal of Molecular Catalysis. B, Enzymatic, 33(3-6) (33)
Operational stability of subtilisin CLECs in organic solvents in repeated batch and in continuous operation
Fernandes JFA, et al.
Biochemical Engineering Journal, 24(1) (24)
Michaela Kreiner et al.
Biotechnology and bioengineering, 87(1), 24-33 (2004-06-24)
Recently, we reported a new high-activity biocatalyst for use in organic media termed protein-coated microcrystals (PCMC) (Kreiner et al. [2001] Chem Commun 12:1096-1097). These novel particles consist of water-soluble micron-sized crystalline particles coated with the given biocatalyst(s) and are prepared
A study of the intermolecular interactions of tolmetin/N-acetyl-L-tyrosine ethyl ester complex.
Tito A, Jimenez-Lopez C, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 72, 1000-1006 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service