A6710
Albumin hydrolysate
Synonym(s):
Albumin solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.25
Recommended Products
biological source
protein from chicken egg white
form
powder
technique(s)
microbiological culture: suitable
storage temp.
2-8°C
General description
Ovalbumin is the major constituent of egg white protein produced in the hen′s oviduct, particularly in the magnum tissue. This glycoprotein represents about 54% of the total proteins of egg albumin. Ovalbumin is a member of the serine protease inhibitor (SERPIN) family.
Application
Albumin hydrolysate has been used:
- to study its effects on the content and profile of volatile aroma compounds of various food products
- to determine the degree of interaction of free or included form of hydroxycinnamic and chlorogenic acids (CHAs) with egg ovalbumin and to analyze the stability of interactions during proteolysis
- as a supplement in various food types to evaluate their bioaccessibility after food processing as well as after in vitro enzymatic hydrolysis
In a rather unusual application, albumin hydrolysate enhanced ethanol production during fermentation by Saccharomyces sake (sake yeast). In media supplemented with albumin hydrolysate, final alcohol concentration was 60% higher than without supplementation. The effect is believed to be due to enhanced alcohol tolerance of cells grown in supplemented medium.
Biochem/physiol Actions
Ovalbumin contributes to the formation of egg white. It elicits various biological activities, such as antihypertensive, anticancer, antimicrobial, antioxidant, and immune-modulation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Matheus M Pereira et al.
Process biochemistry (Barking, London, England), 51(6), 781-791 (2016-09-20)
The ability of aqueous biphasic systems (ABS) composed of polyethylene glycols of different molecular weights (PEG 400, 600 and 1000) and buffered aqueous solutions of potassium citrate/citric acid (pH = 5.0 - 8.0) to selectively extract ovalbumin from egg white
Mylene Da Silva et al.
Biology of reproduction, 93(3), 71-71 (2015-07-15)
The ovalbumin gene family in Gallus gallus is composed of three homologous genes located within a 46 kb locus on chromosome 2: ovalbumin, ovalbumin-related protein Y (OVAY), and ovalbumin-related protein X (OVAX) genes. The expression of these genes in hen
Effect of addition of green coffee extract and nanoencapsulated chlorogenic acids on aroma of different food products
Budryn G, et al.
LWT--Food Science and Technology, 73, 197-204 (2016)
Interactions of free and encapsulated hydroxycinnamic acids from green coffee with egg ovalbumin, whey and soy protein hydrolysates
Budryn G, et al.
LWT--Food Science and Technology, 65, 823-831 (2016)
Role of Materials Science in Food Bioengineering (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service