Skip to Content
Merck
All Photos(1)

Key Documents

A6512

Sigma-Aldrich

L-Arginine β-naphthylamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21N5O · HCl
CAS Number:
Molecular Weight:
335.83
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

ethanol: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl.N[C@@H](CCCNC(N)=N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C16H21N5O.ClH/c17-14(6-3-9-20-16(18)19)15(22)21-13-8-7-11-4-1-2-5-12(11)10-13;/h1-2,4-5,7-8,10,14H,3,6,9,17H2,(H,21,22)(H4,18,19,20);1H/t14-;/m0./s1

InChI key

WEVOXPYVEJEKIT-UQKRIMTDSA-N

Related Categories

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Ball et al.
Journal of clinical microbiology, 15(1), 28-34 (1982-01-01)
The aminopeptidase activity of arginine-utilizing mycoplasmas was investigated with 20 aminoacyl beta-naphthylamide substrates. High levels of arginyl-beta-naphthylamide hydrolysis were demonstrated in 6 of 11 species when extracts of concentrated washed organisms were used. Relatively low arginine aminopeptidase activity was demonstrated
Yu-Chih Wang et al.
Veterinary microbiology, 142(3-4), 309-312 (2009-11-17)
Enrofloxacin (ER) resistant Actinobacillus pleuropneumoniae strains emerged in Taiwan in 2002. The mechanism of ER resistance in A. pleuropneumoniae has not yet been reported. A total of 48 A. pleuropneumoniae isolates were obtained from the lungs of pigs with pleuropneumonia
S Kawata et al.
Journal of biochemistry, 88(6), 1601-1605 (1980-12-01)
Porcine liver aminopeptidase B[EC 3.4.11.6] is highly specific for hydrolysis of beta-naphthylamides of basic L-amino acids; the Km values for L-arginine beta-naphthylamide and L-lysine beta-naphthylamide were 0.035 and 0.12 mM, respectively. The enzyme was inhibited by various alpha-amino acids. Among
Koo-Youn Kim et al.
International journal of food microbiology, 146(1), 52-56 (2011-03-01)
Sixteen Salmonella strains resistant to nalidixic acid isolated from kimbab, the most popular ready-to-eat (RTE) food in Korea, and chicken meat were selected for this study. The resistant strains were shown to have high minimal inhibitory concentrations (MICs) against nalidixic
G Arechaga et al.
Cellular & molecular biology research, 41(5), 369-375 (1995-01-01)
The subcellular distribution of soluble and membrane-bound Arg-beta-naphthylamide-hydrolyzing activities was studied in the left and right rat brain during development and aging. During development, the soluble activity was heterogeneous, whereas adult animals showed the highest activity in the synaptosomal fraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service