Skip to Content
Merck

Skip To

A6270

Azure A chloride

Dye content ≥70 %, Powder or crystals

Synonym(s):

Azure eosin methylene blue; 7-Amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
EC Number:
208-510-7
MDL number:
Colour Index Number:
52005
Beilstein/REAXYS Number:
3922287
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Azure A chloride, Dye content ≥70 %

form

powder or crystals

Quality Segment

composition

Dye content, ≥70%

color

green to very dark green

mp

290 °C (dec.) (lit.)

solubility

water: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

NC1=CC(SC(C2=N3)=CC(C=C2)=[N+](C)C)=C3C=C1.[Cl-]

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Application

Azure A chloride has been used for the staining of polyacrylamide gel to analyse the bovine lung heparin digestion by heparinase I. It has been used for the determination of the sulphated groups content in polysaccharides.

Biochem/physiol Actions

Azure A is a basic cationic dye. The combination of azure A and silver staining is used for the quantification of glycosaminoglycans in agarose gels. It is also used for the staining of chromosomes in the nucleus. It is a main component of the azure B/polychromed methylene blue – eosin stains family, also referred to as the Romanowsky stain.

Disclaimer

Handle and store under argon. Glove bag item.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
861049A2918200212
color

green to very dark green

color

blue to very dark blue, very dark green

color

dark brown, green to very dark green

color

green to black

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

solubility

water: 1 mg/mL

solubility

H2O: 1 mg/mL

solubility

water: 1 mg/mL

solubility

-

form

powder or crystals

form

powder

form

powder

form

powder or crystals

composition

Dye content, ≥70%

composition

Dye content, 70%

composition

Dye content, ≥70%

composition

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


Still not finding the right product?

Explore all of our products under Azure A chloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters