Skip to Content
Merck
All Photos(2)

Key Documents

A6104

Sigma-Aldrich

Anti-ARP2 antibody, Mouse monoclonal

clone FMS96, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FMS96, monoclonal

form

buffered aqueous solution

mol wt

antigen 40-45 kDa

species reactivity

mouse, human, rat, canine

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total cell extract of C2 mouse myoblast cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTR2(10097)
mouse ... Actr2(66713)
rat ... Actr2(289820)

General description

ARP2 is part of a complex with 7 subunits called ARP2/3. ARP2 is an actin-related protein that function to regulate the actin cytoskeleton.
Anti-ARP2 antibody, Mouse monoclonal, (mouse IgG1 isotype) is derived from the FMS96 hybridoma produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with recombinant human ARP2.
Mouse Monoclonal Anti-ARP2 antibody binds to human, canine, rat, and mouse ARP2, 40-45kDa.

Immunogen

recombinant human ARP2

Application

Anti-ARP2 antibody, Mouse monoclonal has been used in:
  • Immunoblotting
  • Immunocytochemistry
  • Enzyme linked immuosorbent assay

Biochem/physiol Actions

Actin-related protein 2 (ARP2) is a part of ARP2/3 complex and play a key role in actin filament formation. It mediates changes in the polymerization state of actin filaments. The Wiskott-Aldrich Syndrome protein (WASP) regulates ARP2/3 complex of proteins. The ARP2/3 complex and its activator spinocerebellar ataxia, autosomal recessive 2 (Scar2) are involved in Golgi polarization in NIH 3T3 cells. However in primary astrocytes, Golgi polarization involves neither actin cytoskeleton or Arp2/3 complex nor any WASP-family of proteins. In a different model, it was found that the ARP2/3 complex was necessary for neutrophil chemotaxis and phagocytosis. By genetic and loss of function studies, it was demonstrated that Scar is the major regulator of the ARP2/3 complex rather than the WASP family of proteins, the latter having more restricted roles in specific cellular events.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The state of the filament.
Aguda, AH.
The Embo Journal, 6, 220-226 (2005)
Phosphorylation of actin-related protein 2 (Arp2) is required for normal development and cAMP chemotaxis in Dictyostelium
Choi CH, et al.
The Journal of biological chemistry, 288(4) (2013)
The WASP--Arp2/3 pathway: genetic insights
Vartiainen MK and Machesky LM
Current Opinion in Cell Biology, 16(2) (2004)
Involvement of the Arp2/3 complex and Scar2 in Golgi polarity in scratch wound models
Magdalena J, et al.
Molecular Biology of the Cell, 14(2), 670-684 (2003)
SCAR is a primary regulator of Arp2/3-dependent morphological events in Drosophila
Zallen JA, et al.
The Journal of cell biology, 156(4) (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service