Skip to Content

A6043

Albumin, biotinamidocaproyl labeled bovine

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥80% protein basis (biuret)
Form:
lyophilized powder
Technique(s):
bioconjugation: suitable
Concentration:
>80% protein (biuret)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥80% protein basis (biuret)

form

lyophilized powder

origin

USA origin

concentration

>80% protein (biuret)

extent of labeling

≥8 mol biotin per mol albumin

technique(s)

bioconjugation: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Application

Albumin-biotin conjugate was used to develop a sensitive non-isotopic benzodiazepine receptor assay. The method can be generalized to other receptors.

Physical form

Lyophilized powder containing sodium citrate buffer salts

Preparation Note

Prepared from albumin (A7638)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library