Skip to Content
Merck

Skip To

A6003

Bovine Serum Albumin

lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.25
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥96% (agarose gel electrophoresis), ≥96%
Form:
lyophilized powder
Technique(s):
microbiological culture: suitable
Impurities:
≤0.01% fatty acid (GC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥96% (agarose gel electrophoresis), ≥96%

form

lyophilized powder

purified by

cold ethanol fractionation

packaging

poly bottle of

origin

USA origin

free fatty acid content

≤0.01%

technique(s)

microbiological culture: suitable

impurities

≤0.01% fatty acid (GC)

pH

7

solubility

water: soluble (50 mg/ml)

UniProt accession no.

application(s)

hematology

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

BSA is a known carrier of fatty acids (FA). Thus control over the specific FA′s for culture of cell lines is important, as different cell lines can be differentially sensitive to particular fatty acids. Fatty acid-free BSA thus is a useful tool for cell culture experiments where the specific fatty acid content must be tightly controlled, to allow researchers to use particular fatty acids specific to their cell lines. Fatty acid-free albumin also allows for optimal and maximum binding sites for adding specific fatty acids to use in cell culture. The use of FA-free BSA also mitigates concerns about endogenous FA′s potentially in non-FA-free BSA.
Bovine serum albumin (BSA) is a highly abundant plasma protein that assists in the regulation of the colloidal osmotic pressure of blood. BSA is used in many different applications, such as cell culture, immunochemistry, and protein quantitation.

This BSA product is specifically tested for very low FA content / fatty acid free status.

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Cold ethanol fractionated
  • Essentially fatty acid-free

Preparation Note

Often referred to as Cohn fraction V; this product is prepared by a modified method of the Cohn cold ethanol fractionation method.
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0281A2153A3608
description

lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)

description

lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)

description

lyophilized powder, ≥96% (agarose gel electrophoresis)

description

lyophilized powder, ≥96% (agarose gel electrophoresis)

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

origin

USA origin

origin

USA origin

origin

USA origin

origin

New Zealand origin

technique(s)

microbiological culture: suitable

technique(s)

enzyme immunoassay: suitable

technique(s)

ELISA: suitable, blood typing: suitable, microbiological culture: suitable

technique(s)

immunoprecipitation (IP): suitable

assay

≥96% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Hello, Is this BSA (A6003) low endotoxin?

    1 answer
    1. This product is not tested for endotoxin. Please see the link below to review low endotoxin BSA product options:
      https://www.sigmaaldrich.com/search/bovine-serum-albumin-endotoxin?focus=products&page=1&perpage=30&sort=relevance&term=bovine%20serum%20albumin%20endotoxin&type=product

      Helpful?

  2. What is the BSA used in I2771?

    1 answer
    1. The same BSA used in I2771 is A6003. It is FAA free and has no lipid profile.

      Helpful?

Reviews

No rating value

Active Filters