Skip to Content
Merck
All Photos(1)

Key Documents

A5384

Sigma-Aldrich

Aconitase from porcine heart

Synonym(s):

Aconitate hydratase, Citrate (isocitrate) hydrolyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Porcine heart

Quality Level

form

solid

mol wt

~66 kDa by gel filtration

storage temp.

−20°C

Application

Aconitase may be usefull in diabetes research as well as to study structural conservation between iron-responsive element binding proteins (IRE-BP) and isomerases .

Biochem/physiol Actions

Aconitase catalyses the stereo-specific isomerization of citrate to isocitrate via cis-aconitate in the tricarboxylic acid cycle. Aconitase is inhibited by cyanide, sulfide and copper and mercury at low concentrations. It is competitively inhibited by trans-aconitate. Aconitase contains an internal Fe-S cluster which is responsible for its activity.

Unit Definition

One unit will convert 1.0 μmole of citrate (via cis-aconitate) to isocitrate per min at pH 7.4 at 25 °C.

Analysis Note

Crude preparation containing aconitase activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey F Kelso et al.
Chemistry & biology, 19(10), 1237-1246 (2012-10-30)
Superoxide (O(2)(·-)) is the proximal mitochondrial reactive oxygen species underlying pathology and redox signaling. This central role prioritizes development of a mitochondria-targeted reagent selective for controlling O(2)(·-). We have conjugated a mitochondria-targeting triphenylphosphonium (TPP) cation to a O(2)(·-)-selective pentaaza macrocyclic
S S Popov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(7), 32-35 (2012-10-03)
A combined therapy with melaxen led to a decrease in the activity of gamma-glutamyl transpeptidase and the level of diene conjugates in blood serum of patients with the drug-induced hepatitis developing on the background of administration of antituberculous preparations. These
Megan J Morgan et al.
Plant physiology, 161(1), 397-407 (2012-11-21)
Organic acid content is regarded as one of the most important quality traits of fresh tomato (Solanum lycopersicum). However, the complexity of carboxylic acid metabolism and storage means that it is difficult to predict the best way to engineer altered
Ewelina Michta et al.
Environmental microbiology, 14(12), 3203-3219 (2012-11-03)
In many organisms, aconitases have dual functions; they serve as enzymes in the tricarboxylic acid cycle and as regulators of iron metabolism. In this study we defined the role of the aconitase AcnA in Streptomyces viridochromogenes Tü494, the producer of
Lucy M Hinder et al.
The Journal of endocrinology, 216(1), 1-11 (2012-10-23)
Diabetic neuropathy (DN) is the most common complication of diabetes and is characterized by distal-to-proximal loss of peripheral nerve axons. The idea of tissue-specific pathological alterations in energy metabolism in diabetic complications-prone tissues is emerging. Altered nerve metabolism in type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service