Skip to Content
Merck
All Photos(3)

Key Documents

A5285

Sigma-Aldrich

Adenosine 5′-diphosphate monopotassium salt dihydrate

bacterial, ≥95%, powder

Synonym(s):

Adenosine 5′-Diphosphate Potassium Salt, ADP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14KN5O10P2 · 2H2O
CAS Number:
Molecular Weight:
501.32
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.77

biological source

bacterial

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[O-]P(OP(OC[C@H]([C@@H](O)[C@H]1O)O[C@H]1N2C3=C(N=C2)C(N)=NC=N3)(O)=O)(O)=O.[K+]

InChI

1S/C10H15N5O10P2.K/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1

InChI key

ZNCWUOPIJTUALR-MCDZGGTQSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine diphosphate (ADP) is a crucial component for energy flow in living organisms. Its structure consists of a sugar backbone connected to adenine and two phosphate groups attached to the fifth carbon atom of the ribose sugar. ADP serves a precursor for ATP biosynthesis. ADP acts as a weak platelet agonist, specifically targeting the P2Y receptor. However, ADP plays a vital role in platelet function by enhancing the effects of other platelet agonists and promoting the stability of platelet aggregates when released from platelet dense granules.

Application

Adenosine 5′-diphosphate has been used to study the release, neuronal effects, and removal of extracellular β-nicotinamide adenine dinucleotide (β-NAD+) in the rat brain. Adenosine 5′-diphosphate has also been used to compare prasugrel versus clopidogrel antiplatelet therapy after acute coronary syndrome.
Adenosine 5′-diphosphate monopotassium salt dihydrate (ADP) has been used:
  • in titration to determine complex I-supported respiration in permeabilized muscle fibers
  • in titrations to determine ADP-stimulated respiratory kinetics
  • in the preparation of ADP solution for measuring mitochondrial respiration

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Platelet P2 Receptors
Cattaneo M
Platelets (2013)
Roles of AMP, ADP, ATP, and AMPK in Healthy Energy Boosting and Prolonged Life Span
Yadav D , et al.
Chemistry & Biology (2017)
Daniel S Ziemianowicz et al.
Journal of the American Society for Mass Spectrometry, 31(2), 207-216 (2020-02-08)
The functional properties of a protein are strongly influenced by its topography, or the solvent-facing contour map of its surface. Together with crosslinking, covalent labeling mass spectrometry (CL-MS) has the potential to contribute topographical data through the measurement of surface
Metabolic remodeling of dystrophic skeletal muscle reveals biological roles for dystrophin and utrophin in adaptation and plasticity
Hardee JP, et al.
Molecular Metabolism (2021)
Leonie Durnin et al.
The European journal of neuroscience, 35(3), 423-435 (2012-01-27)
Recent evidence supports an emerging role of β-nicotinamide adenine dinucleotide (β-NAD(+) ) as a novel neurotransmitter and neuromodulator in the peripheral nervous system -β-NAD(+) is released in nerve-smooth muscle preparations and adrenal chromaffin cells in a manner characteristic of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service