Skip to Content
Merck

A5253

Albumin from chicken egg white

powder, 62-88% (agarose gel electrophoresis)

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
eCl@ss:
42010201
EC Number:
232-692-7
NACRES:
NA.25
MDL number:
Biological source:
chicken egg white
Form:
powder
Technique(s):
bioconjugation: suitable
Concentration:
62-88% (agarose gel electrophoresis)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken egg white

Quality Segment

type

Grade II

form

powder

mol wt

44287 Da by calculation

concentration

62-88% (agarose gel electrophoresis)

technique(s)

bioconjugation: suitable

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

General description

Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.
Ovalbumin make 75% of the total egg white protein.

Application

A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.
Albumin from chicken egg white has also been used to investigate the effects of infusion fluids (colostrum whey, milk whey, amniotic fluid) on ovalbumin (OVA) absorption in fetal pig intestine.

Biochem/physiol Actions

Differential scanning calorimetry indicates that the chicken egg albumin denatures at 84 °C. Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen.
Albumin from chicken egg white serves as a natural adhesive.

Analysis Note

Crude dried egg white


Still not finding the right product?

Explore all of our products under Albumin from chicken egg white


pictograms

Health hazard

signalword

Danger

hcodes

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

Hazard Classifications

Resp. Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library