Skip to Content
Merck

A4551

α-Amylase from Bacillus licheniformis

lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)

Sustainability features

Synonym(s):

1,4-α-D-Glucan-glucanohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-560-9
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
500-1,500 units/mg protein
Assay:
93-100% (SDS-PAGE)
Biological source:
Bacillus licheniformis
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Bacillus licheniformis

Quality Segment

assay

93-100% (SDS-PAGE)

form

lyophilized powder

specific activity

500-1,500 units/mg protein

mol wt

58-62 kDa

composition

Protein, 60-80% BCA

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Sustainability features

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Physical form

Lyophilized powder containing potassium phosphate

Other Notes

One unit will liberate 1.0 mg of maltose from starch in 3 min at pH 6.9 at 20 °C.


Still not finding the right product?


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library