Skip to Content
Merck

A4378

Bovine Serum Albumin

lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥97% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
ELISA: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥97% (agarose gel electrophoresis)

form

lyophilized powder

quality

crystallized

purified by

cold ethanol fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

ELISA: suitable

pH

7

solubility

water: soluble (50 mg/ml)

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

Albumin is the most abundant plasma protein in humans. High levels of albumin are associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Bovine Serum Albumin has been:
  • as an additive in washout solution and fluorescent dye solution
  • as a supplement in Waymouth medium and modified essential medium-α (MEMα )
  • in plant extract for the protein precipitation test
  • as a component in Ringer′s solution to rinse the retinal tissue

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Often referred to as Cohn fraction V; this product is prepared by a modified method of the Cohn cold ethanol fractionation method. Further purified by a proprietary crystallization method
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library