Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
alkaline phosphatase conjugate
Clone:
polyclonal
Application:
ELISA (d), IHC (p), WB
Citations:
67
biological source
rabbit
Quality Segment
conjugate
alkaline phosphatase conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous glycerol solution
species reactivity
goat
technique(s)
direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
General description
IgG antibodies regulate several functions such as complement activation and phagocytosis. Thus they play a crucial role in facilitating cytological immune responses. Polyclonal anti-goat IgG (whole molecule)–alkaline phosphatase antibody (diluted 1:15,000) can be used as a secondary antibody for immunoblotting of Sall4. This antibody can also be used in immunohistochemistry (diluted1:400) . Rabbit anti-goat IgG antibody reacts specifically with goat IgG and normal goat serum.
Primary goat antibodies are often used to study target proteins for various clinical and research purposes. Thus, secondary anti-goat antibody conjugated to a detectable substrate can be used to facilitate the accurate detection and localization of target proteins.
Immunogen
Purified goat IgG.
Application
Anti-Goat IgG (whole molecule)-Alkaline Phosphatase antibody is suitable for use in immunoblotting. The product can also be used for direct ELISA (1:30,000) and immunohistochemistry (1:50 using formalin-fixed, paraffin-embedded sections).
Detection of TLR2 and TLR4 in protein extracts from epidermal keratinocytes was performed by western blot using alkaline phosphatase conjugated rabbit anti-goat IgG as the secondary at a 1:2500 dilution.
Biochem/physiol Actions
Binds all goat Igs
Physical form
Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A4187-.5ML | 04061837514319 |
| A4187-.25ML | 04061837514302 |
| A4187-.05ML | 04061835805068 |
| A4187-1ML | 04061837514326 |