Skip to Content
Merck

A3765

N-(p-Aminophenyl)oxamic acid–Agarose

saline suspension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

saline suspension

Quality Segment

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

20 atoms

capacity

≥37 units/mL binding capacity (neuraminidase (N2876, 4MU-NANA substrate))

storage temp.

2-8°C

SMILES string

[X]Nc1ccc(cc1)NC(=O)C(=O)O

Application

N-(p-Aminophenyl)oxamic acid agarose is used in affinity chromatography, protein chromatography and in specialty resins. N-(p-Aminophenyl)oxamic acid has been used in studies characterizing sialidase, neuraminidase, β-N-acetylhexosaminidases and β-galactosidase.

Physical form

Suspension in 0.1 M NaCl containing preservative


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library