Skip to Content
Merck

Skip To

A3699

Acrylamide/Bis-acrylamide solution

30%, BioReagent, suitable for electrophoresis, 37.5:1

Acrylamide/Bis-acrylamide, 30% solution

Synonym(s):

30% Acrylamide/Bisacrylamide 37.5:1, Polyacrylamide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acrylamide/Bis-acrylamide, 30% solution, BioReagent, suitable for electrophoresis, 37.5:1

sterility

0.2 μm filtered

Quality Segment

product line

BioReagent

form

liquid

feed ratio

37.5:1

technique(s)

electrophoresis: suitable

suitability

suitable for electrophoresis

storage temp.

2-8°C

SMILES string

NC(=O)C=C.C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2.C3H5NO/c1-3-6(10)8-5-9-7(11)4-2;1-2-3(4)5/h3-4H,1-2,5H2,(H,8,10)(H,9,11);2H,1H2,(H2,4,5)

InChI key

OCQZXMCGTAWGEQ-UHFFFAOYSA-N

General description

Polyacrylamide gels are prepared using acrylamide and bis-acrylamide, where bis-acrylamide is a cross-linking agent. On the other hand, acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.

Application

These 30% (w/w) acrylamide/bis-acrylamide solutions are available at the most common ratios for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
Acrylamide/Bis-acrylamide, 30% solution has been used:
  • in the preparation of polyacrylamide gel for photobleaching experiment
  • as a constituent in the preparation of gel precursor solution and pore-gradient microgel precursor solution
  • in the preparation of polyacrylamide gels during western blot analyses

Preparation Note

Solution prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3574A3449A9926
form

liquid

form

liquid

form

liquid

form

liquid

feed ratio

37.5:1

feed ratio

29:1

feed ratio

19:1

feed ratio

19:1

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

suitability

suitable for electrophoresis

suitability

suitable for electrophoresis

suitability

suitable for electrophoresis

suitability

suitable for electrophoresis

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review