Skip to Content
Merck
All Photos(1)

Key Documents

A3313

Sigma-Aldrich

Anti-Human Polyvalent Immunoglobulins (α, γ and μ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Human immunoglobulins are glycoproteins that regulate immune responses to infections and allergies. Anti-human polyvalent immunoglobulins can be used for serological analyses in diseased population groups. These antibodies may also be used for studying xenotransplantation models and immunological disorders such as rheumatoid arthritis. Anti-Human Polyvalent Immunoglobulins (α, γ and μ-chain specific)-Alkaline Phosphatase antibody is specific for human IgG, IgA and IgM when tested against human IgA, IgG, IgM, Bence Jones κ, and λ myeloma proteins.

Immunogen

Purified human IgG, IgA and IgM

Application

Anti-Human Polyvalent Immunoglobulins (α, γ and μ-chain specific)-Alkaline Phosphatase antibody is suitable for use in western blot and immunostaining techniques.
The presence of IgG was detected in normal human sera using alkaline phosphatase-conjugated goat anti-human polyvalent IgG (chain specific) by ELISA at a concentration of 1:1000. An ELISA assay was performed using alkaline phosphatase-conjugated goat anti-human IgG polyvalent antibody to detect natural antibodies to KLH in human sera.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 10 mM glycine, 1 mM MgCl2, 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John S Spencer et al.
Clinical and vaccine immunology : CVI, 18(2), 260-267 (2010-12-24)
A simple serodiagnostic test based on the Mycobacterium leprae-specific phenolic glycolipid I(PGL-I), for individuals with leprosy is nearly universally positive in leprosy patients with high bacillary loads but cannot be used as a stand-alone diagnostic test for the entire spectrum
Angela Barone et al.
The Journal of biological chemistry, 288(14), 10035-10050 (2013-02-14)
Due to their pluripotency and growth capability, there are great expectations for human embryonic stem cells, both as a resource for functional studies of early human development and as a renewable source of cells for use in regenerative medicine and
Luisa Fernanda Duarte et al.
Colombia medica (Cali, Colombia), 45(2), 61-66 (2014-08-08)
To compare the diagnostic performance of seven methods to determine Trypanosoma cruzi infection in patients with chronic Chagas disease. Analytical study, using the case-control design, which included 205 people (patients with Chagasic cardiomyopathy, n=100; control group, n=105). Three enzyme linked
Stefan Magnusson et al.
Xenotransplantation, 12(1), 49-58 (2004-12-16)
The use of thin easily revascularized cutaneous nerve autografts, which has been the gold standard, or the alternative use of nerve allografts or artificial grafts for nerve reconstructing have all their pros and cons. Nerve xenotransplantation may offer a potential
Béatrice Volney et al.
Acta tropica, 82(1), 11-23 (2002-03-21)
This paper describes a sero-epidemiological study of malaria prevalence in French Guiana. An immunofluorescence assay and an enzyme-linked immunosorbent assay were used to detect antibodies against blood-stage antigens and synthetic peptides mimicking the repetitive epitope of the sporozoites of Plasmodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service