Skip to Content
Merck

Skip To

A3264

Albumin from sheep serum

lyophilized powder

Synonym(s):

Adh1p

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
sheep
Assay:
≥97% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
hybridization: suitable, microbiological culture: suitable, protein quantification: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

assay

≥97% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

hybridization: suitable, microbiological culture: suitable, protein quantification: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

sheep ... ALB(443393)

General description

Albumin is the most abundant plasma protein in humans. High levels of albumin are associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Albumin from sheep serum has been used in the quantitative determination of different albumin species by albumin blue580 fluorescence assay. It has also been used in perfusion solutions.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2764A1830A4538
technique(s)

hybridization: suitable, protein quantification: suitable, microbiological culture: suitable

technique(s)

ELISA: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

biological source

sheep

biological source

Porcine

biological source

Porcine

biological source

rat

assay

≥97% (agarose gel electrophoresis)

assay

96-99% (agarose gel electrophoresis)

assay

≥98% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters