Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.77
EC Number:
232-570-3
MDL number:
Specific activity:
≥100 units/mg protein
Biological source:
bovine liver
biological source
bovine liver
Quality Segment
form
powder
specific activity
≥100 units/mg protein
composition
Protein, ≥70% biuret
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
cow ... ARG2(518752)
General description
L-arginase is also called as L-arginine amidinohydrolase. It exists in two forms, such as arginase-1 and arginase-2. Arginase-1 is present in liver cells and arginase-2 is seen usually in extrahepatic tissues like, kidney, brain, skeletal muscle, small intestine and the lactating mammary gland. Arginase -2 is mapped to human chromosome 14q24.1−24.3.
Biochem/physiol Actions
L-arginase hydrolyze L-arginine into L-ornithine and urea, which is the last step of the urea cycle in the liver of ureotelic species. Arginase plays a major role in the mammalian immune system and the enzyme participates in several aspects of inflammation.
L-Arginase is the major degradative enzyme for arginine; converts arginine to ornithine.
L-Arginase is the major degradative enzyme for arginine; converts arginine to ornithine; deficiency is associated with spasticity and motor dysfunction.
Other Notes
One unit will cause the hydrolysis of 1.0 μmole of L-arginine per minute at pH 9.5 and 37 °C.
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
