Skip to Content
Merck

A2861

Astemizole

≥98% (HPLC), hERG potassium channel blocker, powder

Synonym(s):

1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H31FN4O
CAS Number:
Molecular Weight:
458.57
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41121800
EC Number:
272-441-9
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Astemizole, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

storage condition

desiccated, protect from light

solubility

DMSO: >20 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

COc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1

InChI

1S/C28H31FN4O/c1-34-25-12-8-21(9-13-25)14-17-32-18-15-24(16-19-32)30-28-31-26-4-2-3-5-27(26)33(28)20-22-6-10-23(29)11-7-22/h2-13,24H,14-20H2,1H3,(H,30,31)

InChI key

GXDALQBWZGODGZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Biochem/physiol Actions

Astermizole is a potent hERG potassium channel blocker (IC50 of 0.9 nM) and may used as a pharmacological chaperone to correct folding defects and restore protein function for some mutated forms of hERG channels. It has also been studied for treatment of malaria, hERG and hEAG channel function in cancer and as a second generation antihistamine H-1 antagonist.
Astermizole is a potent hERG potassium channel blocker (IC50 of 0.9 nM).

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Astemizole


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library