Skip to Content
Merck
All Photos(3)

Key Documents

A2767

Sigma-Aldrich

Adenosine 5′-triphosphate–Agarose

lyophilized powder

Synonym(s):

5′-ATP agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
eCl@ss:
32160414
NACRES:
NA.56

biological source

plant (Sea weed)

Quality Level

form

lyophilized powder

extent of labeling

1-5 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

C-8

matrix spacer

9 atoms

storage temp.

−20°C

SMILES string

[X]Nc1ncnc2[n](cnc21)[C@@H]3O[C@@H]([C@H]([C@H]3O)O)CO[P](=O)(O[P](=O)(O[P](=O)(O)O)O)O

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Adenosine 5′-triphosphate–Agarose (5′-ATP-agarose) is a conjugate of 5′-ATP to crosslinked 4% beaded agarose (activated by cyanogen bromide), via the C-8 atom of 5′-ATP. 5′-ATP–agarose is applicable for affinity purification of various proteins and enzymes like cyclin-dependent kinase 2 (CDK2), heat shock proteins, and cryptochromes.

Application

Adenosine 5′-triphosphate Agarose (5′-ATP agarose) has been used in affinity chromatography to purify uridine kinase from Ehrlich ascites tumor cells.
Adenosine 5′-triphosphate–Agarose (5′-ATP–agarose) is applicable for affinity purification of various proteins and enzymes like cyclin-dependent kinase 2 (CDK2), heat shock proteins-70 (HSP-70), and cryptochromes. 5′-ATP agarose has been used in affinity chromatography to purify uridine kinase from Ehrlich ascites tumor cells.

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J E Braun et al.
The Journal of biological chemistry, 271(42), 25989-25993 (1996-10-18)
Cysteine string protein (CSP) is a 34 kDa secretory vesicle protein bearing a "J-domain" as well as a palmitoylated cysteine-rich "string" region used for membrane attachment. Mutation of the CSP gene causes impaired presynaptic neuromuscular transmission in Drosophila melanogaster, implicating
E T Bui et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(18), 9651-9656 (1996-09-03)
Trichomonads are among the earliest eukaryotes to diverge from the main line of eukaryotic descent. Keeping with their ancient nature, these facultative anaerobic protists lack two "hallmark" organelles found in most eukaryotes: mitochondria and peroxisomes. Trichomonads do, however, contain an
Lutz Fischer et al.
British journal of pharmacology, 142(5), 861-868 (2004-06-16)
5-Lipoxygenase (5-LO) is a crucial enzyme in the synthesis of the bioactive leukotrienes (LTs) from arachidonic acid (AA), and inhibitors of 5-LO are thought to prevent the untowarded pathophysiological effects of LTs. In this study, we present the molecular pharmacological
Su Ying Wu et al.
Structure (London, England : 1993), 11(4), 399-410 (2003-04-08)
A family of 4-heteroaryl-2-amino-pyrimidine CDK2 inhibitor lead compounds was discovered with the new database-mining program LIDAEUS through in silico screening. Four compounds with IC(50) values ranging from 17 to 0.9 microM were selected for X-ray crystal analysis. Two distinct binding
Christian Feisst et al.
The Journal of pharmacology and experimental therapeutics, 315(1), 389-396 (2005-07-15)
Myrtucommulone (MC) and semimyrtucommulone (S-MC) are unique oligomeric, nonprenylated acylphloroglucinols contained in the leaves of myrtle (Myrtus communis). Although extracts of myrtle have been traditionally used in folk medicine for the treatment of various disorders, studies addressing select cellular or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service