Skip to Content
Merck
All Photos(1)

Documents

A1929

Sigma-Aldrich

Anti-Monkey IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Monkey IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

baboon, monkey, marmoset

technique(s)

direct ELISA: 1:40,000
dot blot: 1:60,000 (direct chemiluminescence assay)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG (immunoglobulin G) antibody subtype is a predominant serum immunoglobulin of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. IgG is found in four subclasses IgG1, IgG2, IgG3 and IgG4. IgG has a monomeric structure.
IgGs are glycoprotein antibodies that modulate several immune responses. Serum IgG levels of rhesus monkeys are comparable to that of men, hence these monkeys are often used as models for studying immunological disorders. Thus, anti-monkey IgGs can be useful tools for the detection and localization of monkey IgGs. Anti-Monkey IgG (whole molecule)-Alkaline Phosphatase antibody binds to monkey, baboon and marmoset IgGs.

Immunogen

Purified monkey IgG (rhesus)

Application

Alkaline phosphatase-conjugated rabbit anti-monkey IgG antibody has been used as a secondary antibody in enzyme linked immunsorbent assay (ELISA) to detect the level of IgG in serum from treated marmoset monkeys. It has also been used for western blot analysis of to identify maternal IgGs in serum from Rhesus monkeys after lentiviral gene transfer.

Biochem/physiol Actions

IgG antibody participates in complement fixation and opsonization.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Challacombe et al.
Immunology, 36(2), 331-338 (1979-02-01)
Rhesus monkey IgG, IgA and IgM were purified from pooled normal serum and labelled with 125I. Four monkeys were injected intravenously with approximately 50 microgram of each purified immunoglobulin containing between 25 muCi and 50 muCi of 125I. Iodination was
L Boon et al.
Journal of immunology (Baltimore, Md. : 1950), 167(5), 2942-2949 (2001-08-18)
Inhibition of CD40-CD40 ligand interaction is a potentially effective approach for treatment of autoimmune diseases, such as multiple sclerosis. We have investigated this concept with a chimeric antagonist anti-human CD40 mAb (ch5D12) in the marmoset monkey experimental autoimmune encephalomyelitis (EAE)
Stefano Colloca et al.
Science translational medicine, 4(115), 115ra2-115ra2 (2012-01-06)
Replication-defective adenovirus vectors based on human serotype 5 (Ad5) induce protective immune responses against diverse pathogens and cancer in animal models, as well as elicit robust and sustained cellular immunity in humans. However, most humans have neutralizing antibodies to Ad5
Alexander D Douglas et al.
Nature communications, 10(1), 1953-1953 (2019-04-28)
Malaria vaccine design and prioritization has been hindered by the lack of a mechanistic correlate of protection. We previously demonstrated a strong association between protection and merozoite-neutralizing antibody responses following vaccination of non-human primates against Plasmodium falciparum reticulocyte binding protein
IL-21-induced isotype switching to IgG and IgA by human naive B cells is differentially regulated by IL-4
Avery DT, et al.
Journal of Immunology, 181(3), 1767-1779 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service