Skip to Content
Merck

A166

Sp-Adenosine 3′,5′-cyclic monophosphorothioate triethylammonium salt hydrate

≥98% (HPLC), solid

Synonym(s):

Sp-Cyclic 3′,5′-hydrogen phosphorothioate adenosine hydrate, Sp-cAMPS triethylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H11N5O5PS · C6H16N · xH2O
CAS Number:
Molecular Weight:
446.46 (anhydrous basis)
MDL number:
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106305
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D −25.43°, c = 0.99 in H2O(lit.)

color

white

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

CCN(CC)CC.Nc1ncnc2n(cnc12)[C@@H]3O[C@@H]4CO[P@@](S)(=O)O[C@H]4[C@H]3O

InChI

1S/C10H12N5O5PS.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7-4(19-10)1-18-21(17,22)20-7;1-4-7(5-2)6-3/h2-4,6-7,10,16H,1H2,(H,17,22)(H2,11,12,13);4-6H2,1-3H3/t4-,6-,7-,10-,21+;/m1./s1

InChI key

OXIPZMKSNMRTIV-BIQAKYJPSA-N

Biochem/physiol Actions

A potent, membrane-permeable activator of cAMP-dependent protein kinase I and II.
Sp-Diastereomer of adenosine 3′,5′-cyclic monophosphorothioate is a potent, membrane-permeable activator of cAMP dependent protein kinase I and II that mimics the effects of cAMP as a second messenger in numerous systems while being resistant to cyclic nucleotide phosphodiesterases; exhibits greater specificity and affinity than forskolin and cAMP analogs such as dibutyryl-cAMP.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Yusta et al.
Journal of neurochemistry, 51(6), 1808-1818 (1988-12-01)
We have compared the effects of forskolin, N6,2'-O-dibutyryladenosine 3':5'-cyclic monophosphate (dibutyryl cyclic AMP, Bt2-cAMP), and butyrate on several aspects of neuroblastoma cell physiology. The morphology of Neuro 2A cells was similar after incubation with forskolin and Bt2-cAMP, which caused extensive
H G Scholübbers et al.
European journal of biochemistry, 138(1), 101-109 (1984-01-02)
The ability of 24 systematically modified analogues of adenosine 3',5'-monophosphate (cAMP) to enhance the synthesis of beta-galactosidase in glucose-repressed Escherichia coli strains KNBL 1001 and cpd- Crookes has been investigated. The properties of the analogues in comparison with cAMP are
John M E Nichols et al.
Developmental cell, 48(4), 491-505 (2019-01-08)
Signaling from chemoattractant receptors activates the cytoskeleton of crawling cells for chemotaxis. We show using phosphoproteomics that different chemoattractants cause phosphorylation of the same core set of around 80 proteins in Dictyostelium cells. Strikingly, the majority of these are phosphorylated

Global Trade Item Number

SKUGTIN
A166-5MG04061832895628
A166-10MG04061832895611
A166-1MG04061833342725

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service