Skip to Content
Merck

A143

(S)-(−)-Atenolol

powder

Synonym(s):

(−)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHNHCH2CH(OH)CH2OC6H4CH2CONH2
CAS Number:
Molecular Weight:
266.34
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

optical activity

[α]23/D −10.5°, c = 0.5 in H2O(lit.)

color

white to beige

mp

148-152 °C (lit.)

solubility

0.1 M HCl: soluble

SMILES string

CC(C)NC[C@H](O)COc1ccc(CC(N)=O)cc1

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)/t12-/m0/s1

InChI key

METKIMKYRPQLGS-LBPRGKRZSA-N

Gene Information

human ... ADRB1(153)

Application

(S)-(-)-Atenolol may be used as a standard in the preparation of enantiomer for high-performance liquid chromatography-tandem mass spectrometry studies (HPLC-MS/MS). It has also been used in the β blocking molecule for Raman spectroscopy studies.

Biochem/physiol Actions

(S)-(-)-Atenolol is a β1-adrenergic receptor antagonist and an active enantiomer of atenolol.


Still not finding the right product?

Explore all of our products under (S)-(−)-Atenolol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library