Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H23N5O14P2
CAS Number:
Molecular Weight:
559.32
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
MDL number:
Assay:
≥93%
Biological source:
yeast (candida utilis)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless
Storage temp.:
−20°C
Product Name
Adenosine 5′-diphosphoribose sodium salt, ≥93%
biological source
yeast (candida utilis)
Quality Segment
assay
≥93%
form
powder
solubility
water: 50 mg/mL, clear, colorless
storage temp.
−20°C
SMILES string
[Na].Nc1ncnc2n(cnc12)C3OC(COP(O)(=O)OP(O)(=O)OCC(O)C(O)C(O)C=O)C(O)C3O
InChI
1S/C15H23N5O14P2.Na.H/c16-13-9-14(18-4-17-13)20(5-19-9)15-12(26)11(25)8(33-15)3-32-36(29,30)34-35(27,28)31-2-7(23)10(24)6(22)1-21;;/h1,4-8,10-12,15,22-26H,2-3H2,(H,27,28)(H,29,30)(H2,16,17,18);;
InChI key
NWPVXDZLQTURKI-UHFFFAOYSA-N
Application
5′-Deoxy-5′-(methylthio)adenosine has been used:
- to give a range of nicotinamide adenine dinucleotide (NADH)
- oxidation rates for the respiratory complex (CI/CIII/CIV)
- CI/alternative(AOX) pathway
- to measure poly(ADP-ribose) glycohydrolaseactivity
- poly(ADP-ribose) (PAR) binding competition assay
- oxidase (AOX) pathway
- to measure poly(ADP-ribose) glycohydrolase activity
- poly(ADP-ribose) (PAR) binding competition assay
Biochem/physiol Actions
Adenosine5′-diphosphoribose (ADP-Ribose) is used as the basic building block used as a substrate in the formation of poly(ADP-ribose) by members of the poly(ADP-ribose) polymerase (PARP) family. Poly(ADP-ribose) formation and degradation are important components of DNA repair and apoptosis signaling. ADP-Ribose initiates the transient receptor potential subfamily melastatin, type2 (TRPM2), a Na+, Ca2+, and K+ cation channel.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
