Skip to Content
Merck

A0639

Albumin from rabbit serum

lyophilized powder, ≥96% (agarose gel electrophoresis)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
rabbit
Assay:
≥96% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
ELISA: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

ELISA: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

rabbit ... ALB(100009195)



Still not finding the right product?

Explore all of our products under Albumin from rabbit serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library