Skip to Content

A0295

Anti-Human IgA (α-chain specific)−Peroxidase antibody produced in goat

affinity isolated antibody

Synonym(s):

Goat Anti-Human IgA (α-chain specific)−HRP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
peroxidase conjugate
Clone:
polyclonal
Application:
DB, ELISA (d), IHC (p), WB
Citations:
71
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

4-11 mg/mL

technique(s)

direct ELISA: 1:50,000, dot blot: 1:80,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100, western blot: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human IgA (a-chain specific)-peroxidase antibody is isolated from goat anti-human IgA antiserum by immunospecific purification to remove essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to the alpha chain of human IgA.Goat antihuman IgA is conjugated to peroxidase and purified to remove unconjugated material. Specificity of the peroxidase conjugated anti-human IgA is determined by immunoelectrophoresis (IEP) and cross-reactivity of the conjugate is determined by enzyme linked immunoassays (ELISA).

Immunogen

Purified human IgA.

Application

Anti-Human IgA (α-chain specific) Peroxidase antibody produced in goat has been used in:
  • Enzyme linked immunosorbent assays (ELISA) at dilution of 1:3,000 and 1: 1,600
  • Tissue anti-transglutaminase antibody (tTG) assay at a dilution of 1:1,000
  • Dot blot: a minimum working dilution of 1:80,000 was determined in a direct chemiluminescence assay using 10ng human IgA/dot. Luminol plus enhancer was used as substrate.
  • Immunohistology using formalin-fixed, paraffin-embedded sections at a dilution of 1:100
  • Transcytosis assay of dimeric IgA (D-IgA)
  • Western Blotting

Biochem/physiol Actions

Anti-Human IgA (a-chain specific)-Peroxidase antibody produced in goat conjugate is specific for human IgA when tested against human IgA, IgG, IgM, Bence Jones Kappa and Lambda myeloma proteins. The antibody conjugate does not react with mouse or rat IgG.
IgA is a secretory glycoprotein antibody that regulates the physiological balance between commensal intestinal bacteria and the defenses of the host immune system. It is the highest generated antibody isotype. IgAs are mainly secreted in the intestinal mucosa and prevent the commensal microorganisms from invading the intestines.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT as preservative

Preparation Note

For continuous use, store at 2-8°C for a maximum of one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.
Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library