Skip to Content
Merck

A0262

Lithium potassium acetyl phosphate

≥85% (elemental analysis), powder

Synonym(s):

Acetyl phosphate lithium potassium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3COOP(O)(OK)(OLi)
CAS Number:
Molecular Weight:
184.06
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
304-378-0
MDL number:
Beilstein/REAXYS Number:
5689718
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Lithium potassium acetyl phosphate, high-energy phosphate donor

Quality Segment

assay

≥85% (elemental analysis)

form

powder

mp

>300 °C (lit.)

solubility

water: 25 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li+].[K+].CC(=O)OP([O-])([O-])=O

InChI

1S/C2H5O5P.K.Li/c1-2(3)7-8(4,5)6;;/h1H3,(H2,4,5,6);;/q;2*+1/p-2

InChI key

RLQMPLKXFIXRCV-UHFFFAOYSA-L

Application

Lithium potassium acetyl phosphate:
  • has been used as a substrate for CpxR (transcription factor) in x-ray crystallography to soak CpxR receiver domain (CpxRRD) crystals to obtain the phosphorylated form
  • may be used as a phosphate (P) donor in a study to screen a wide range of natural and synthetic organic phosphate donors with several enzymes on various hydroxyl?compounds for the improved enzymatic synthesis of phosphate monoesters
  • has been used to set up premixA for in vitro translation reaction

Biochem/physiol Actions

Lithium potassium acetyl phosphate is commonly used as the source for acetyl phosphate. So acetyl phosphate can be used as a suitable alternative to replace inorganic oligophosphates in phosphatase-catalyzed transphosphorylation to reduce inorganic monophosphate by-product formation.


Still not finding the right product?

Explore all of our products under Lithium potassium acetyl phosphate


pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library