Skip to Content
Merck

A0232

Alfuzosin hydrochloride

≥98% (HPLC), solid

Synonym(s):

N-(3-((4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino)propyl)tetrahydro-2-furancarboxamide hydrochloride, Uroxatral hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H27N5O4 · HCl
CAS Number:
Molecular Weight:
425.91
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: >10 mg/mL, H2O: insoluble

originator

Sanofi Aventis

SMILES string

Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)C3CCCO3

InChI

1S/C19H27N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h10-11,14H,4-9H2,1-3H3,(H,21,25)(H2,20,22,23);1H

InChI key

YTNKWDJILNVLGX-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Alfuzosin hydrochloride is an alpha-adrenergic blocker used to treat benign prostatic hyperplasia (BPH). It works by relaxing the muscles in the prostate and bladder neck, making it easier to urinate.
Alfuzosin hydrochloride is an alpha-adrenergic blocker.

Features and Benefits

This compound is featured on the α1-Adrenoceptors and α2-Adrenoceptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Alfuzosin hydrochloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jarosław Szulfer et al.
Journal of chromatography. A, 1346, 69-77 (2014-05-06)
This paper focuses on the application of a column classification system based on the Katholieke Universiteit Leuven for the characterization of physicochemical properties of core-shell and ultra-high performance liquid chromatographic stationary phases, followed by the verification of the reliability of
Satyanarayan Pattnaik et al.
Medicina (Kaunas, Lithuania), 51(4), 253-261 (2015-10-02)
There has been relatively little investigation of the effect of temperature on skin permeation compared to other methods of penetration enhancement. A principal physicochemical factor which controls the passive diffusion of a solute from a vehicle into the skin arises



Global Trade Item Number

SKUGTIN
A0232-5MG04061832071473
A0232-25MG04061832071466