Skip to Content
Merck

Skip To

96404

Atto 550-Streptavidin

BioReagent, suitable for fluorescence

Synonym(s):

Streptavidin-Atto 550

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12352209
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 554 nm; λem 576 nm in 0.1 M phosphate pH 7.0

λ

in PBS (buffer pH 7.4)

suitability

corresponds for gel electrophoresis, suitable for fluorescence

storage temp.

−20°C

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. These labels have rigid structures that do not show any cis-trans-isomerization. They display exceptional intensity with minimal spectral shift on conjugation.

Analysis Note

free of unconjugated dye

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
563044070949937
suitability

corresponds for gel electrophoresis, suitable for fluorescence

suitability

corresponds for gel electrophoresis, suitable for fluorescence

suitability

-

suitability

corresponds for gel electrophoresis, suitable for fluorescence

form

solid

form

-

form

powder

form

solid

fluorescence

λex 554 nm; λem 576 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 565 nm; λem 590 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 590 nm; λem 619 nm in 0.1 M phosphate pH 7.0

fluorescence

-

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters