Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
human lung
Quality Segment
description
Human bronchial epithelium, normal
form
liquid
manufacturer/tradename
ECACC
packaging
tube of 5 μg 95102433-DNA-5UG, pkg of vial of cells 95102433-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
Application
Biochem/physiol Actions
CSF1PO: 9,12
D13S317: 13
D16S539: 12
D5S818: 12,13
D7S820: 10,13
THO1: 7,9.3
TPOX: 6,11
vWA: 17,18
Packaging
Preparation Note
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line.
Please view the Terms & Conditions of Supply for more information.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 10092312, human lung, bronchoalveolar, epithelial-like. Thick stellate cells with vacuoles. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 10092311, human lung, Epithelial-like: cells are spindle-shaped with few vacuoles | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. |
| morphology Epithelial-like | morphology Epithelial-like: Thick stellate shaped cells with vacuoles. | morphology Epithelial-like: cells are spindle-shaped with few vacuoles | morphology Epithelial-like: cells are spindle-shaped with few vacuoles. Upon confluence develop an epithelioid "cobblestone-like mat" |
| karyotype Not specified | karyotype Not specified | karyotype Not specified | karyotype Not specified |
| biological source human lung | biological source human lung (bronchoalveolar) | biological source human lung | biological source human lung |
| growth mode Adherent | growth mode Adherent | growth mode Adherent | growth mode Adherent |
| receptors Not specified | receptors Not specified | receptors Not specified | receptors Not specified |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

