Skip to Content
Merck

94498

5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride

≥85% (HPLC)

Synonym(s):

5-Cyano-2,3-bis(4-methylphenyl)-2H-tetrazolium chloride, CTC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H14ClN5
CAS Number:
Molecular Weight:
311.77
UNSPSC Code:
12171500
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8954451
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥85% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

[Cl-].Cc1ccc(cc1)-n2nc(n[n+]2-c3ccc(C)cc3)C#N

InChI

1S/C16H14N5.ClH/c1-12-3-7-14(8-4-12)20-18-16(11-17)19-21(20)15-9-5-13(2)6-10-15;/h3-10H,1-2H3;1H/q+1;/p-1

InChI key

SSJZAXOTLCJNLF-UHFFFAOYSA-M

General description

5-cyano-2,3-di-(p-tolyl)-tetrazolium chloride is a Redox Sensitive tetrazolium salt producing a fluorescent formazan (only in solid state) upon reduction; It indicates the respiratory activity in cells.

Application

5-cyano-2,3-di(p-tolyl) tetrazolium chloride (CTC)-reduction test is used for live/dead analysis. 5-cyano-2,3-di-(p-tolyl)-tetrazolium chloride has been employed in histochemistry and for determining cell viability, for eukaryotic cell viability assays and bacterial viability experiments, 5-cyano-2,3-di-(p-tolyl)-tetrazolium chloride (CTC) is the most often employed compound.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library