Skip to Content
Merck

94417

(+)-Magnesium L-ascorbate

≥90% (T)

Synonym(s):

L(+)-Ascorbic acid magnesium salt, Vitamin C magnesium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14MgO12
CAS Number:
Molecular Weight:
374.54
UNSPSC Code:
12352205
NACRES:
NA.79
PubChem Substance ID:
EC Number:
239-442-6
Beilstein/REAXYS Number:
3767527
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (T)

form

powder

color

white to faintly beige

SMILES string

OC[C@@H](O)[C@@H]1OC(=O)C(O)=C1O[Mg]OC2=C(O)C(=O)O[C@H]2[C@H](O)CO

InChI

1S/2C6H8O6.Mg/c2*7-1-2(8)5-3(9)4(10)6(11)12-5;/h2*2,5,7-10H,1H2;/q;;+2/p-2/t2*2-,5+;/m11./s1

InChI key

AIOKQVJVNPDJKA-GVKKNTFRSA-L

General description

(+)-Magnesium L-ascorbate or vitamin C magnesium salt is a water-soluble molecule. It is naturally present in a variety of foods such as citrus fruits and vegetables.

Biochem/physiol Actions

L-Ascorbic acid (Vitamin C) exhibits anti-oxidant properties in both plants and animals. In plants, it facilitates various cellular processes such as growth, programmed cell death, pathogen responses, hormone responses, flowering and senescence, as well as protection against environmental stresses. Ascorbic acid acts as a reducing agent in various enzymatic reactions to reduce oxidative stress. L-Ascorbate has an ability to inhibit bacterial mutagenesis induced by N-nitroso compounds. Vitamin C deficiency leads to scurvy, prominent-gingivitis, perifollicular hemorrhages, poor wound healing, petechiae and ecchymosis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library