Skip to Content
Merck
All Photos(1)

Key Documents

94060901

FTC-133 Cell Line human

Synonym(s):

FTC133 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human thyroid (lymph node metastasis)

Quality Level

description

Human follicular thyroid carcinoma

form

liquid

growth mode

Adherent

karyotype

Complex chromosomal changes

morphology

Polymorphic

products

Expression of 5′ - Deiodinase Type I

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human follicular thyroid carcinoma, lymph node metastasis

Cell Line Description

FTC-133 was obtained from a lymph node metastasis of a follicular thyroid carcinoma not the primary tumour as originally reported from a 42-year-old male. The morphology differs from flat polygonal to spindle formed cells. They retained differentiated thyrocyte function and thyrocyte responsiveness to thyrotropin and local active growth factors. Thyroglobulin immunoreactivity could be shown in the cytoplasm and EGF receptor immunoreactivity could be visualised on the cell membranes. Complex chromosomal changes were detected as well as a p53 mutation. FTC-133 (Sigma catalogue number 94060901), FTC-236 (Sigma catalogue number 06030202) and FTC-238 (Sigma catalogue number 94060902) were all derived from the same individual, a 42 year old male with follicular thyroid cancer. This has been confirmed by short tandem repeat STR-PCR analysis at ECACC The Y chromosome could not be detected in these cell line by (STR)-PCR analysis. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell lines are identical to the source provided by the depositor based on the STR-PCR analysis.

Application

FTC-133 cell line has been used to study the function of human thyroid and development of thyroid cancer.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11
D16S539: 11
D5S818: 12
D7S820: 9,10
THO1: 9.3
TPOX: 9
vWA: 15,18

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 to 1:12 i.e. seeding at 1-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hoelting et al.
Biochemical and biophysical research communications, 195(3), 1230-1236 (1993-09-30)
The signal transduction of TSH in invasion and growth of FTC 133, a human follicular thyroid cancer cell line, was investigated. TSH (0.01-1 mIU/ml) stimulated invasion of FTC 133 by 21% and growth by 20% of basal. Cyclic AMP-stimulators and
Wanfeng Yu et al.
Human molecular genetics, 24(1), 142-153 (2014-08-26)
Germline mutations in the PTEN tumor-suppressor gene and germline variations in succinate dehydrogenase subunit D gene (SDHD-G12S, SDHD-H50R) are associated with a subset of Cowden syndrome and Cowden syndrome-like individuals (CS/CSL) and confer high risk of breast, thyroid and other
Hypermethylation of a New Distal Sodium/Iodide Symporter (NIS) Enhancer (NDE) Is Associated With Reduced NIS Expression in Thyroid Tumors
Ana Luiza Galrao
The Journal of Clinical Endocrinology and Metabolism, 99(6) (2014)
T Hölting et al.
European journal of endocrinology, 132(2), 229-235 (1995-02-01)
We have shown recently that epidermal growth factor (EGF) enhanced invasion and growth of differentiated thyroid cancer cells in vitro and in vivo. The present study analyzed the effects of transforming growth factor alpha (TGF-alpha) on invasion and growth of
Thomas A Werner et al.
Scientific reports, 7(1), 11383-11383 (2017-09-14)
Follicular thyroid carcinoma's (FTC) overall good prognosis deteriorates if the tumour fails to retain radioactive iodine. Therefore, new druggable targets are in high demand for this subset of patients. Here, we investigated the prognostic and biological role of survivin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service