Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H14N2O4
CAS Number:
Molecular Weight:
190.20
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Product Name
DL-2,6-Diaminopimelic acid, ≥95% (TLC)
assay
≥95% (TLC)
form
powder
SMILES string
OC([C@@H](N)CCC[C@H](N)C(O)=O)=O
InChI
1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m0/s1
InChI key
GMKMEZVLHJARHF-WHFBIAKZSA-N
Application
DL-2,6-Diaminopimelic acid, a stereoisomer of meso-DAP, may be used as a reference material in assays used to identify markers of bacteria and fungi in the rumen of ruminants.
Biochem/physiol Actions
Stereoisomer of meso-DAP. meso-DAP is a biosynthetic precursor of the essential amino acid L-lysine and component of peptidoglycan in the cell wall of many bacteria. As these functions are not observed in mammalian biochemistry, alpha,alpha′-diamino diacids (DADs) are of interest as potential antibiotics with low mammalian toxicity.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Analysis Note
diastereometric purity ≥98.0% (HPLC)
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 92591-10MG | 04061833269961 |
| 92591-50MG | 04061832405544 |
