Skip to Content
Merck

Skip To

92283

Polymyxin B solution

20 mg/mL in H2O

Synonym(s):

Aerosporin, Polymyxin B, Polymyxin B Sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
51283106
NACRES:
NA.85
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Bacillus polymyxa (fermentation)

Quality Segment

form

liquid

optical activity

[α]/D 20/D (Rotation: -2.3 +/- 0.2 degrees)

concentration

20 mg/mL in H2O

color

colorless to slightly yellow

solubility

H2O: soluble 20 mg/mL

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, fungi

mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N1CCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1=O)CCO)CCN)CCN)CC(C)C)Cc2ccccc2)CCN)NC(=O)C(NC(=O)C(NC(=O)C(NC=O)CCN)CCO)CCN

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

General description

Polymyxin B sulfate is a strongly cationic cyclic polypeptide antibiotic that is derived from fermentation of Bacilus polymyxa. It is a mixture of B1 and B2 sulfate. The product has been used clinically to treat infections of the urinary tract, meninges and blood stream caused by susceptible strains of Pseudomonas aeruginosa.

Application

Polymyxin B is used to study multidrug-resistant pathogens, as an immobilized agent for removal of endotoxins, and to induce pore formation in the membranes of cortex cells from excised sorghum roots.

Biochem/physiol Actions

Mode of Action: Polymyxin B Sulfate binds to the lipid A portion of bacterial lipopolysaccharides, disrupting the cytoplasmic membrane by inducing pores large enough to permit nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane.

Mode of Resistance: The activity of Polymyxin B sulfate is inhibited by iron(II), Co(II), Mn(II) and Magnesium ions. Polymyxin B may also be incompaitible with other microbial agents, including amphoterecin, cephalothin sodium, cephasolin sodium and heparin sodium.

Antimicrobial Spectrum: Has bactericidal action on most gram-negative bacilli, including E. Coli and on most fungi and gram-positive bacteria.

Packaging

10ml, 125ml

Preparation Note

This product is concentrated at 20 mg/mL in water, and should be stored at 2-8°C.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Handle and store under inert gas.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1547007PHR1595P4932
mode of action

cell membrane | interferes

mode of action

-

mode of action

-

mode of action

cell membrane | interferes

antibiotic activity spectrum

Gram-negative bacteria, fungi, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

Gram-negative bacteria

Quality Level

200

Quality Level

-

Quality Level

300

Quality Level

200

form

liquid

form

powder

form

-

form

powder

concentration

20 mg/mL in H2O

concentration

-

concentration

-

concentration

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Polymyxin B solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is this used for eye problems

    1 answer
    1. Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.

      Helpful?

Reviews

No rating value

Active Filters