Skip to Content
Merck

916

α-Naphthyl Acetate Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
41116124
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

12.5 mg/mL

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC(=O)Oc1cccc2ccccc12

InChI

1S/C12H10O2/c1-9(13)14-12-8-4-6-10-5-2-3-7-11(10)12/h2-8H,1H3

InChI key

VGKONPUVOVVNSU-UHFFFAOYSA-N

Application

Reagent used in the 91A procedure for the demonstration of non-specific esterase activity in leukocytes.

Other Notes

12.5 mg/mL, in methanol solution with stabilizers


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

98.1 °F - closed cup

flash_point_c

36.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
669997-100MG04061833554791
669997-500MG04061833554807
534342000104054839117121
700174P-5MG04061838472427
Z720798-20EA04061832967301
Z720836-85EA04061832967332
T2386-100MG04061838256430
T2386-25MG04061838065445
T-047-1ML04061837330094
OP181-100UG04055977208856
Z720771-75EA04061834388296
Z687162-100EA04061832967011
Z720801-20EA04061834388302
T4764-1ML04061837346712
39382-10MG04061834390497
D6315-1CS04061834379935
D6440-1CS04061834379942
D6789-1CS04061826744420
D7039-1CS04061834374145
H9649-1MG04061826675267
T-032-1ML04061837328978
C4045-5MG04061832628738
C3920-5MG04061832628707