Skip to Content
Merck

Skip To

88315

Thesit®

Synonym(s):

Polyethylene glycol dodecyl ether, Polyethylene glycol 400 dodecyl ether, Hydroxypolyethoxydodecane, Lauromacrogol, Macrogol lauryl ether, Polidocanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HO(CH2CH2O)n(CH2)11CH3
CAS Number:
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

form

solid

mol wt

average mol wt 583

impurities

≤1 ppm peroxides (as H2O2)

CMC

0.1 mM (20-25°C)

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Application

Suggested substitute for Lubrol in the preparation of G protein from bovine brain.
Thesit has been used in the reaction buffer to reduce the background sedimentation.

Biochem/physiol Actions

Thesit/Polidocanol possesses in vitro and in vivo anti-angiogenic effects. This sclerosant is mainly used for sclerotherapy.

Legal Information

Thesit is a registered trademark of Desitin Arzneimittel GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
STS021176437P8925
description

-

description

-

description

BioXtra, ≥98.0% (GC)

description

≥98% (GC)

form

solid

form

solid

form

semisolid

form

solid

impurities

≤1 ppm peroxides (as H2O2)

impurities

≤3.0% water

impurities

-

impurities

-

CMC

0.1 mM (20-25°C)

CMC

91 μM

CMC

-

CMC

0.11

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

mol wt

average mol wt 583

mol wt

-

mol wt

406.60 g/mol

mol wt

average mol wt 66,000

description

non-ionic

description

non-ionic

description

non-ionic

description

non-ionic


Signal Word

Warning

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves

Pictograms

Exclamation mark

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters