Skip to Content
Merck

87918

Tetramethylrhodamine isothiocyanate mixed isomers

suitable for fluorescence, mixture of isomers

Synonym(s):

MRITC, TRITC, Tetramethylrhodamine B isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H21N3O3S
CAS Number:
Molecular Weight:
443.52
MDL number:
NACRES:
NA.32
UNSPSC Code:
12352116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

solubility

DMSO: soluble, methanol: soluble

fluorescence

λex 529 nm; λem 596 nm in DMSO

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Tetramethylrhodamine isothiocyanate, also known as TRITC is a fluorescent antibody labelling agent. It is a red-fluorescing marker and can be tracked through fluorescence microscopy.

TRITC dissolves completely in DMSO at a rapid rate. And gamma globulin is unaffected to up to 30% DMSO dilution in carbonate buffer. Tetramethylrhodamine isothiocyanate doesn’t dissolve in other denaturing agents completely.

Application

Tetramethylrhodamine isothiocyanate or TRITC is used for tracking the pathway of calcium phosphate/DNA nanoparticles during cell transfection. It is an extremely intensive fluorochrome used for labeling antibodies.

Other Notes

An extremely intensive fluorochrome for labeling antibodies


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library