Skip to Content

86139

Docusate sodium salt

BioUltra, ≥99.0% (TLC)

Synonym(s):

AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, Dioctyl sulfosuccinate sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H37NaO7S
CAS Number:
Molecular Weight:
444.56
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
209-406-4
Beilstein/REAXYS Number:
4117588
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

product line

BioUltra

assay

≥99.0% (TLC)

form

sheet

mol wt

444.56 g/mol

impurities

insoluble matter, passes filter test

loss

≤1% loss on drying, 110 °C (HV)

solubility

methanol: 0.1 M at 20 °C, clear, colorless

density

1,146 g/cm3 at 27,4 °C

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤100 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

0.1 M in methanol

UV absorption

λ: 260 nm Amax: 0.1, λ: 280 nm Amax: 0.05

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Application

Forms reverse micelles in hydrocarbon solvents; Suitable for the solubilization of the major myelin transmembrane proteolipid


Still not finding the right product?

Explore all of our products under Docusate sodium salt


hcodes

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library