Skip to Content
Merck

85641

Starch dyed with Remazol brilliant blue R

amylase substrate

Synonym(s):

RBB-Starch, Remazol brilliant blue R dyed starch, Starch Azure

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

suitability

suitable as substrate for α-amylase test

Other Notes

Substrate for α-amylase


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A new method for the determination of alpha-amylase.
H Rinderknecht et al.
Experientia, 23(10), 805-805 (1967-10-15)



Global Trade Item Number

SKUGTIN
85641-25G04061833054635