Skip to Content
Merck

854C

Citrate Concentrated Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
12352106
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Citrate solutions are suitable for use in absence of microbial growth.

IVD

for in vitro diagnostic use

concentration

1.5 M

pH

2.9-3.1 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

Application

pH approx. 3.0 when diluted 1:50. For the preparation of a 60% citrate buffered acetone solution used as a fixative for blood smears.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 1

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library