Skip to Content
Merck

83465

RBS T230

concentrate

Synonym(s):

RBS Viro

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161900
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

foaming detergent

Quality Segment

form

liquid

quality

concentrate

technique(s)

HPLC: suitable, protein quantification: suitable

pH

6.0-7.5

density

1.07 g/mL at 20 °C

Application

foaming detergent for cleaning all glass and metallic equipment in virology and medicine

Other Notes

Sales restrictions may apply

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library