Skip to Content
Merck
All Photos(1)

Documents

83388

Sigma-Aldrich

Quercetin 3-D-galactoside

≥97.0% (HPLC)

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-D-galactoside, Hyperin, Hyperoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
5784795
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.47

Assay

≥97.0% (HPLC)

form

powder

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15+,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-DTGCRPNFSA-N

Gene Information

mouse ... Hexa(15211)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Protects against peroxide-induced oxidative damage to cells by scavenging reactive oxygen species and enhancing activity of anti-oxidant enzymes, in particular, catalase and glutathione peroxidase.

Other Notes

A flavonol glycoside widely distribute in plants, including mimosa and St. John′s wort.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sae-Kwang Ku et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 53, 197-204 (2012-12-12)
The potential anticoagulant activities of two single compounds, isorhamnetin-3-O-galactoside (IMG) and hyperoside, from Oenanthe javanica, were tested. The anticoagulant activities were investigated by measuring activated partial thromboplastin time (aPTT) and prothrombin time (PT), and the ability to inhibit production of
Rui-Li Liu et al.
Brain research, 1469, 164-173 (2012-07-10)
Hyperoside is a flavonoid compound and widely used in clinic to relieve pain and improve cardiovascular functions. However, the effects of hyperoside on ischemic neurons and the molecular mechanisms remain unclear. Here, we used an in vitro ischemic model of
Zi-lin Li et al.
Free radical biology & medicine, 57, 132-140 (2013-01-08)
One of the leading causes of death in the world is ischemia/reperfusion (I/R)-mediated acute myocardial infarction. There are a lot of Chinese traditional patent medicines, such as Xin'an capsules, Xin Xuening tablets, and so on, which have protective effects against
Zi-lin Li et al.
Journal of ethnopharmacology, 139(2), 388-394 (2011-11-29)
Hyperoside (Hyp) is a flavonoid compound isolated from Rhododendron ponticum L. leaves that elicits vascular protective effects in vitro. Treatment with Hyp has been found to attenuate endothelial cell damage induced by oxidative stress, but its mechanisms of action remain
Joshua M Hicks et al.
Journal of AOAC International, 95(5), 1406-1411 (2012-11-28)
A single-laboratory-validated NMR spectroscopy method was established for determining the quantity of chlorogenic acid and hyperoside from crude extract material of blueberry leaves of the species Vaccinium angustifolium var. laevifolium House. The calibration curve of chlorogenic acid showed a highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service